ATC311762297 Atlantic Research Chemicals Ltd

3-Chloro-4-[(3-fluorobenzyl)oxy]aniline

  • MDL number MFCD06809822
  • Cas Number : 202197-26-0
  • Molecular Weight : 251.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE001167
Availability details for
Quantity
  • Availability
  • Add to Cart