ATC311762308 Atlantic Research Chemicals Ltd

6-Methoxy-1,1'-biphenyl-3-amine hydrochloride

  • MDL number MFCD01630736
  • Cas Number : 92028-21-2
  • Molecular Weight : 235.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE001178
Availability details for
Quantity
  • Availability
  • Add to Cart