ATC311762326 Atlantic Research Chemicals Ltd

5-Chloro-2-nitro-4-(trifluoromethyl)aniline

  • MDL number MFCD01312277
  • Cas Number : 35375-74-7
  • Molecular Weight : 240.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE001195
Availability details for
Quantity
  • Availability
  • Add to Cart