ATC311762334 Atlantic Research Chemicals Ltd

4-(3-Aminophenyl)pyridine

  • MDL number MFCD00956758
  • Cas Number : 40034-44-4
  • Molecular Weight : 170.214

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=AE00126
Availability details for
Quantity
  • Availability
  • Add to Cart