ATC311762336 Atlantic Research Chemicals Ltd

2-(3-Aminophenyl)pyridine dihydrochloride

  • MDL number MFCD08235170
  • Cas Number : 1170936-92-1
  • Molecular Weight : 243.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE00128
Availability details for
Quantity
  • Availability
  • Add to Cart