ATC311762359 Atlantic Research Chemicals Ltd

2-Chloro-4,6-dimethoxyaniline

  • MDL number MFCD01567107
  • Cas Number : 82485-84-5
  • Molecular Weight : 187.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE00156
Availability details for
Quantity
  • Availability
  • Add to Cart