ATC311762370 Atlantic Research Chemicals Ltd

N-(3,4-Dichlorophenyl)-N-methylamine

  • MDL number MFCD00052850
  • Cas Number : 40750-59-2
  • Molecular Weight : 176.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE00167
Availability details for
Quantity
  • Availability
  • Add to Cart