ATC311762429 Atlantic Research Chemicals Ltd

4-Pyrrolidin-1-ylpyridin-2-amine

  • MDL number MFCD08062967
  • Cas Number : 722550-01-8
  • Molecular Weight : 163.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE002117
Availability details for
Quantity
  • Availability
  • Add to Cart