ATC311762435 Atlantic Research Chemicals Ltd

5-Bromo-6-methylpyridin-3-amine

  • MDL number MFCD08060462
  • Cas Number : 186593-43-1
  • Molecular Weight : 187.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE002122
Availability details for
Quantity
  • Availability
  • Add to Cart