ATC311762454 Atlantic Research Chemicals Ltd

tert-Butyl 6-Chloropyridin-3-ylcarbamate

  • MDL number MFCD08062421
  • Cas Number : 171178-45-3
  • Molecular Weight : 228.678

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=AE00214
Availability details for
Quantity
  • Availability
  • Add to Cart