ATC311762463 Atlantic Research Chemicals Ltd

tert-Butyl 4-[(5-Aminopyridin-2-yl)oxy]piperidine-1-carboxylate

  • MDL number MFCD12026334
  • Cas Number : 346665-41-6
  • Molecular Weight : 293.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE002148
Availability details for
Quantity
  • Availability
  • Add to Cart