ATC311762491 Atlantic Research Chemicals Ltd

tert-Butyl 6-Bromopyridin-2-ylcarbamate

  • MDL number MFCD07367929
  • Cas Number : 344331-90-4
  • Molecular Weight : 273.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE002173
Availability details for
Quantity
  • Availability
  • Add to Cart