ATC311762522 Atlantic Research Chemicals Ltd

2-(4-Fluorophenoxy)pyridin-3-amine

  • MDL number MFCD00067803
  • Cas Number : 175135-64-5
  • Molecular Weight : 204.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE002201
Availability details for
Quantity
  • Availability
  • Add to Cart