ATC311762526 Atlantic Research Chemicals Ltd

2-Chloro-5-fluoropyridin-4-amine

  • MDL number MFCD16622225
  • Cas Number : 89510-90-7
  • Molecular Weight : 146.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE002205
Availability details for
Quantity
  • Availability
  • Add to Cart