ATC311762539 Atlantic Research Chemicals Ltd

3-Amino-4-phenylpyridine

  • MDL number MFCD04114255
  • Cas Number : 146140-99-0
  • Molecular Weight : 170.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE00229
Availability details for
Quantity
  • Availability
  • Add to Cart