ATC311762542 Atlantic Research Chemicals Ltd

1-(3-Aminopyridin-4-yl)ethanone

  • MDL number MFCD07375078
  • Cas Number : 13210-52-1
  • Molecular Weight : 136.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE00232
Availability details for
Quantity
  • Availability
  • Add to Cart