ATC311762571 Atlantic Research Chemicals Ltd

2-Bromo-5-chloropyridin-3-amine

  • MDL number MFCD07368976
  • Cas Number : 90902-83-3
  • Molecular Weight : 207.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE00262
Availability details for
Quantity
  • Availability
  • Add to Cart