ATC311762578 Atlantic Research Chemicals Ltd

N-Allylpyridin-3-amine

  • MDL number MFCD09033527
  • Cas Number : 204773-12-6
  • Molecular Weight : 134.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE00269
Availability details for
Quantity
  • Availability
  • Add to Cart