ATC311762653 Atlantic Research Chemicals Ltd

1H-Pyrazol-5-amine

  • MDL number MFCD08700632
  • Cas Number : 916420-28-5
  • Molecular Weight : 83.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE00911
Availability details for
Quantity
  • Availability
  • Add to Cart