ATC311762688 Atlantic Research Chemicals Ltd

3-Methoxypyrazin-2-amine

  • MDL number MFCD05722793
  • Cas Number : 4774-10-1
  • Molecular Weight : 125.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE01105
Availability details for
Quantity
  • Availability
  • Add to Cart