ATC311762692 Atlantic Research Chemicals Ltd

6-Amino-2-thioxo-2,3-dihydropyrimidin-4(1H)-one

  • MDL number MFCD00052384
  • Cas Number : 1004-40-6
  • Molecular Weight : 143.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE01201
Availability details for
Quantity
  • Availability
  • Add to Cart