ATC311762713 Atlantic Research Chemicals Ltd

4-Chloro-6-ethylpyrimidin-2-amine

  • MDL number MFCD09800366
  • Cas Number : 5734-67-8
  • Molecular Weight : 157.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE01222
Availability details for
Quantity
  • Availability
  • Add to Cart