ATC311762727 Atlantic Research Chemicals Ltd

6-Chloro-5-nitropyrimidin-4-amine

  • MDL number MFCD00127772
  • Cas Number : 4316-94-3
  • Molecular Weight : 174.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE01236
Availability details for
Quantity
  • Availability
  • Add to Cart