ATC311762743 Atlantic Research Chemicals Ltd

6-Iodopyridazin-3-amine

  • MDL number MFCD09991754
  • Cas Number : 187973-60-0
  • Molecular Weight : 221

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE01305
Availability details for
Quantity
  • Availability
  • Add to Cart