ATC311762774 Atlantic Research Chemicals Ltd

5-Methyl-1H-benzimidazol-2-amine

  • MDL number MFCD01658297
  • Cas Number : 6285-68-3
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE02202
Availability details for
Quantity
  • Availability
  • Add to Cart