ATC311762817 Atlantic Research Chemicals Ltd

3-Aminopiperidine

  • MDL number MFCD04972497
  • Cas Number : 54012-73-6
  • Molecular Weight : 100.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE03720
Availability details for
Quantity
  • Availability
  • Add to Cart