ATC311762835 Atlantic Research Chemicals Ltd

N,1-Dimethylpyrrolidin-3-amine

  • MDL number MFCD00059061
  • Cas Number : 13005-11-3
  • Molecular Weight : 114.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE04503
Availability details for
Quantity
  • Availability
  • Add to Cart