ATC311762864 Atlantic Research Chemicals Ltd

4,6-Dimethoxy-1,3,5-triazin-2-amine

  • MDL number MFCD00052765
  • Cas Number : 16370-63-1
  • Molecular Weight : 156.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE05007
Availability details for
Quantity
  • Availability
  • Add to Cart