ATC311762904 Atlantic Research Chemicals Ltd

6-Amino-2-benzofuran-1(3H)-one

  • MDL number MFCD00033530
  • Cas Number : 57319-65-0
  • Molecular Weight : 149.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE10007
Availability details for
Quantity
  • Availability
  • Add to Cart