ATC311762920 Atlantic Research Chemicals Ltd

1-Aminoindan-2-ol

  • MDL number MFCD00053472
  • Cas Number : 7480-35-5
  • Molecular Weight : 149.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE10023
Availability details for
Quantity
  • Availability
  • Add to Cart