ATC311762922 Atlantic Research Chemicals Ltd

tert-Butyl 3-Hydroxy-2,3-dihydro-1H-inden-1-ylcarbamate

  • MDL number MFCD09864900
  • Cas Number : 1086378-71-3
  • Molecular Weight : 249.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE10025
Availability details for
Quantity
  • Availability
  • Add to Cart