ATC311763019 Atlantic Research Chemicals Ltd

2-Methoxy-5-(trifluoromethyl)benzaldehyde

  • MDL number MFCD03788528
  • Cas Number : 146539-83-5
  • Molecular Weight : 204.146

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=AH001110
Availability details for
Quantity
  • Availability
  • Add to Cart