ATC311763040 Atlantic Research Chemicals Ltd

3-(2-Hydroxyethoxy)benzaldehyde

  • MDL number MFCD00191448
  • Cas Number : 60345-97-3
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AH001131
Availability details for
Quantity
  • Availability
  • Add to Cart