ATC311763055 Atlantic Research Chemicals Ltd

3-Amino-4-methoxybenzaldehyde

  • MDL number MFCD07787578
  • Cas Number : 351003-10-6
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AH001146
Availability details for
Quantity
  • Availability
  • Add to Cart