ATC311763137 Atlantic Research Chemicals Ltd

(4-Fluorophenyl)acetaldehyde

  • MDL number MFCD02261761
  • Cas Number : 1736-67-0
  • Molecular Weight : 138.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 85
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AH00184
Availability details for
Quantity
  • Availability
  • Add to Cart