ATC311763217 Atlantic Research Chemicals Ltd

N-(2-Bromo-6-fluorobenzyl)-N-methylamine

  • MDL number MFCD18089185
  • Cas Number : 1287218-39-6
  • Molecular Weight : 218.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AM001125
Availability details for
Quantity
  • Availability
  • Add to Cart