ATC311763244 Atlantic Research Chemicals Ltd

3-Formylpyridine-2-carbonitrile

  • MDL number MFCD07367918
  • Cas Number : 131747-66-5
  • Molecular Weight : 132.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AH00267
Availability details for
Quantity
  • Availability
  • Add to Cart