ATC311763287 Atlantic Research Chemicals Ltd

5-Methyl-1,3-thiazole-2-carbaldehyde

  • MDL number MFCD08060733
  • Cas Number : 13838-78-3
  • Molecular Weight : 127.166

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=AH00604
Availability details for
Quantity
  • Availability
  • Add to Cart