ATC311763311 Atlantic Research Chemicals Ltd

3-(Trifluoromethyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD07375188
  • Cas Number : 1001020-14-9
  • Molecular Weight : 164.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AH00913
Availability details for
Quantity
  • Availability
  • Add to Cart