ATC311763363 Atlantic Research Chemicals Ltd

6-Methyl-1H-indole-3-carbaldehyde

  • MDL number MFCD00049347
  • Cas Number : 4771-49-7
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AH01913
Availability details for
Quantity
  • Availability
  • Add to Cart