ATC311763379 Atlantic Research Chemicals Ltd

2-Quinoxalinecarbaldehyde

  • MDL number MFCD00272280
  • Cas Number : 1593-08-4
  • Molecular Weight : 158.1594

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=AH02801
Availability details for
Quantity
  • Availability
  • Add to Cart