ATC311763392 Atlantic Research Chemicals Ltd

1-(Methylsulphonyl)piperidine-4-carbaldehyde

  • MDL number MFCD11582937
  • Cas Number : 241134-35-0
  • Molecular Weight : 191.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AH03708
Availability details for
Quantity
  • Availability
  • Add to Cart