ATC311763433 Atlantic Research Chemicals Ltd

2-Morpholinobenzylamine

  • MDL number MFCD03086183
  • Cas Number : 204078-48-8
  • Molecular Weight : 192.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AM00107
Availability details for
Quantity
  • Availability
  • Add to Cart