ATC311763436 Atlantic Research Chemicals Ltd

N-(3,5-Difluorobenzyl)-N-methylamine

  • MDL number MFCD11212394
  • Cas Number : 90390-28-6
  • Molecular Weight : 157.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AM001100
Availability details for
Quantity
  • Availability
  • Add to Cart