ATC311763438 Atlantic Research Chemicals Ltd

4-(Benzylamino)butan-1-ol

  • MDL number MFCD00191609
  • Cas Number : 59578-63-1
  • Molecular Weight : 179.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AM001102
Availability details for
Quantity
  • Availability
  • Add to Cart