ATC311763448 Atlantic Research Chemicals Ltd

4-[(Methylamino)methyl]benzonitrile

  • MDL number MFCD09738490
  • Cas Number : 34403-48-0
  • Molecular Weight : 146.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AM001112
Availability details for
Quantity
  • Availability
  • Add to Cart