ATC311763556 Atlantic Research Chemicals Ltd

(4-Methylpyridin-3-yl)methylamine

  • MDL number MFCD06213859
  • Cas Number : 1443-42-1
  • Molecular Weight : 122.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AM00212
Availability details for
Quantity
  • Availability
  • Add to Cart