ATC311763571 Atlantic Research Chemicals Ltd

N-Methyl-N-[(4-methylpyridin-2-yl)methyl]amine

  • MDL number MFCD09800577
  • Cas Number : 1060804-82-1
  • Molecular Weight : 136.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AM00227
Availability details for
Quantity
  • Availability
  • Add to Cart