ATC311763575 Atlantic Research Chemicals Ltd

N-Methyl-N-(4-pyridylmethyl)amine

  • MDL number MFCD00066937
  • Cas Number : 6971-44-4
  • Molecular Weight : 122.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AM00231
Availability details for
Quantity
  • Availability
  • Add to Cart