ATC311763645 Atlantic Research Chemicals Ltd

N-Methyl-N-[(2-methyl-1H-indol-3-yl)methyl]amine

  • MDL number MFCD08056294
  • Cas Number : 335032-50-3
  • Molecular Weight : 174.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AM01902
Availability details for
Quantity
  • Availability
  • Add to Cart